missing translation for 'onlineSavingsMsg'
Learn More

Icariin, MedChemExpress

Product Code. p-200097677
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16482336

Brand: Medchem Express HYN0014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC50s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator.

Icariin is a flavonol glycoside. Icariin inhibits PDE5 and PDE4 activities with IC50s of 432 nM and 73.50 μM, respectively. Icariin also is a PPARα activator.

Specifications

Chemical Name or Material Icariin
CAS 489-32-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C33H40O15
Quantity 500 mg
Synonym Ieariline
Solubility Information DMSO : 125 mg/mL (184.73 mM; Need ultrasonic)
SMILES O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=C(C/C=C(C)/C)C(OC(C3=CC=C(OC)C=C3)=C(O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)C5=O)=C5C(O)=C2)[C@@H]1O
Molecular Weight (g/mol) 676.66
Formula Weight 676.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.