missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (B-T1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £400.00
Specifications
| Antigen | ICAM-2/CD102 |
|---|---|
| Clone | B-T1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207972
|
Novus Biologicals
NB100-66624-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18280351
|
Novus Biologicals
NB100-66624 |
0.1 mg |
£400.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| ICAM-2/CD102 | |
| 1.0 mg/mL | |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P13598 | |
| 3384 | |
| ICAM-2 transfected CHO cells | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| B-T1 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| ICAM2 | |
| IgG1 | |
| Affinity Purified | |
| Recognizes the human CD102 cell surface molecule, a 55-65kD glycoprotein also known as ICAM-2. NB100-66624 inhibits cell adhesion and T cell activation and also recognizes soluble ICAM-2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title