missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (B-T1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66624-0.025MG
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:20-1:50, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| P13598 | |
| ICAM2 | |
| ICAM-2 transfected CHO cells | |
| 0.025 mg | |
| Primary | |
| Recognizes the human CD102 cell surface molecule, a 55-65kD glycoprotein also known as ICAM-2. NB100-66624 inhibits cell adhesion and T cell activation and also recognizes soluble ICAM-2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| B-T1 | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3384 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction