missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Ethyl Ester, TRC

Product Code. 30398070
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398070 1 g
30407072 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398070 Supplier Toronto Research Chemicals Supplier No. TRCI1400801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41283-72-1
Molecular Formula C15 H22 O2
Molecular Weight (g/mol) 234.33
Synonym Ibuprofen Ethyl Ester, 2-(4-Isobutylphenyl)propionic acid ethyl ester, Ethyl 2-(4-isobutylphenyl)propionate, Ethyl 2-(p-isobutylphenyl)propionate, Ethyl 4-isobutyl-α-methylphenylacetate, α-Methyl-4-(2-methylpropyl)-benzeneacetic Acid Ethyl Ester
IUPAC Name ethyl 2-[4-(2-methylpropyl)phenyl]propanoate
SMILES CCOC(=O)C(C)c1ccc(CC(C)C)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H22O2/c1-5-17-15(16)12(4)14-8-6-13(7-9-14)10-11(2)3/h6-9,11-12H,5,10H2,1-4H3
Formula Weight 234.162
Chemical Name or Material Ibuprofen Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.