CAS RN 41283-72-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ibuprofen Ethyl Ester, TRC
CAS: 41283-72-1 Molecular Formula: C15 H22 O2 Molecular Weight (g/mol): 234.33 Synonym: Ibuprofen Ethyl Ester,2-(4-Isobutylphenyl)propionic acid ethyl ester,Ethyl 2-(4-isobutylphenyl)propionate,Ethyl 2-(p-isobutylphenyl)propionate,Ethyl 4-isobutyl-α-methylphenylacetate,α-Methyl-4-(2-methylpropyl)-benzeneacetic Acid Ethyl Ester IUPAC Name: ethyl 2-[4-(2-methylpropyl)phenyl]propanoate SMILES: CCOC(=O)C(C)c1ccc(CC(C)C)cc1
2
Ibuprofen Ethyl Ester, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.