missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ IAEDANS (1,5-IAEDANS, 5-((((2-Iodoacetyl)amino)ethyl)amino)Naphthalene-1-Sulfonic Acid)

Product Code. 11594886
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11594886 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11594886 Supplier Molecular Probes™ Supplier No. I14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IAEDANS (1,5-IAEDANS, 5-((((2-Iodoacetyl)amino)ethyl)amino)Naphthalene-1-Sulfonic Acid)

The fluorescence of the thiol-reactive IAEDANS is quite dependent upon environment. Its conjugates frequently respond to ligand binding by undergoing spectral shifts and changes in fluorescence intensity that are determined by the degree of aqueous solvation. Advantages of this reagent include high water solubility above pH 4 and a relatively long fluorescence lifetime (10-15 nanoseconds), making the conjugates useful for fluorescence polarization and rotational studies. In addition, because it has a large Stokes shift and an emission that overlaps with the absorption of fluorescein, Alexa Fluor™ 488, Oregon Green™ dyes and BODIPY™ FL dyes, IAEDANS is an excellent reagent for fluorescence resonance energy transfer (FRET) measurements.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type IAEDANS
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye Naphthalene
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.