Learn More
uvex I-works Spectacles
Product Code. 31910424 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Clear
Unit Size:
Each
Product Code. Lens Tint unitSize
31910424 Clear Each
1 options
Product Code. 31910424 Supplier uvex Supplier No. 9194265
  • ONLINE EXCLUSIVE PRICE
    £11.55
    £10.61 / Pack of 1
    Save £0.94
    8% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
I-works encapsulates the very best of uvex safety eyewear.
  • The lightweight, sporty design and panoramic lens make the eyewear extremely comfortable to wear - even over long periods of time.
  • The outstanding, long-lasting uvex supra-vision excellence coating technology, uvex i-works safety spectacles boast long-lasting anti-fogging properties that withstand repeated cleaning.
  • Comfortable, soft duo-component side arms ensure an easy-to-wear, non-slip fit.
  • Sporty, rugged safety spectacles
  • Available with different lens tints
  • Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
  • Fits the uvex IES Better for the environment, since lenses can be replaced as necessary
  • Metal-free
  • Ergonomically shaped side arms with soft ear-pieces ensure a comfortable, secure fit without creating pressure points
  • Anatomically shaped nose piece for all-day wearer comfort
  • Certified in accordance with EN 166 (personal eye protection) and EN 170 (UV filters) UV400 - 100% protection against harmful UV radiation up to 400 nm
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Anti-scratch, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material TPU
UV Protection UV400
Frame Type Semi-rimless