missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BacLight™ Red Bacterial Stain
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
1000µg
Description
The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detecting and monitoring bacteria. These two dyes are not nucleic acid stains. Bacteria stained with the BacLight Green and BacLight Red bacterial stains exhibit bright green and red fluorescence (absorption/emission ∼480/516 and ∼581/644 nm, respectively), and can be resolved using the appropriate flow cytometric channels or fluorescence microscopy filters. The BacLight bacterial stains are compatible with formaldehyde or alcohol fixation methods.
Specifications
Specifications
| Cell Type | Bacteria |
| Content And Storage | Contains 20 vials of BacLight™ red bacterial stain (50 ug per vial). Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Bacterial Stain |
| Dye Type | Other Label(s) or Dye(s) |
| Emission | 488 |
| Form | Solid |
| Format | Tube(s), Slide(s) |
| Product Line | BacLight |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction