missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BacLight™ Green Bacterial Stain

Product Code. 11524187
Change view
Click to view available options
Quantity:
20 x 50 μg
Unit Size:
1000µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11524187 20 x 50 μg 1000µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11524187 Supplier Invitrogen™ Supplier No. B35000
  • ONLINE EXCLUSIVE PRICE
    £522.00
    £391.65 / 1000µg
    Save £130.35
    24% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

BacLight™ Green Bacterial Stain

The BacLight Green (Cat. no. B-35000) and BacLight Red (Cat. no. B-35001) bacterial stains are fluorescent labeling reagents for detecting and monitoring bacteria. These two dyes are not nucleic acid stains. Bacteria stained with the BacLight Green and BacLight Red bacterial stains exhibit bright green and red fluorescence (absorption/emission ∼480/516 and ∼581/644 nm, respectively), and can be resolved using the appropriate flow cytometric channels or fluorescence microscopy filters. The BacLight bacterial stains are compatible with formaldehyde or alcohol fixation methods.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Bacteria
Content And Storage Contains 20 vials of BacLight™ green bacterial stain (50 ug per vial). Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Bacterial Stain
Dye Type Other Label(s) or Dye(s)
Form Solid
Format Tube(s), Slide(s)
Product Line BacLight
Quantity 20 x 50 μg
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength 480/ 561
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.