Learn More
I-A/I-E Rat anti-Mouse, FITC, Clone: 2G9, BD
Rat Monoclonal Antibody
£87.38 - £384.17
Specifications
| Antigen | I-A/I-E |
|---|---|
| Clone | 2G9 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821559
|
BD Biosciences
562009 |
50 μg |
£90.95 £87.38 / Each Save £3.57 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15877308
|
BD Biosciences
553623 |
0.5 mg |
£401.00 £384.17 / Each Save £16.83 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2G9 antibody reacts with the I-Ad and I-Ed MHC class II alloantigens. It also reacts with transfectants expressing I-Eb and I-Ek and with cells from mice of the H-2p and H-2q haplotypes. This antibody is non-reactive with cells from SJL (H-2s) and NOD (H-2g7) mice. Flow cytometric analysis indicates that the 2G9 and M5/114.15.2 (Cat. No. 556999, for purified mAb) monoclonal antibodies have comparable reactivity on cells from mice with I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek alloantigens.
Specifications
| I-A/I-E | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| IgG2a κ | |
| Affinity Purified |
| 2G9 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.