Learn More
I-A/I-E Rat anti-Mouse, FITC, Clone: 2G9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553623
Description
The 2G9 antibody reacts with the I-Ad and I-Ed MHC class II alloantigens. It also reacts with transfectants expressing I-Eb and I-Ek and with cells from mice of the H-2p and H-2q haplotypes. This antibody is non-reactive with cells from SJL (H-2s) and NOD (H-2g7) mice. Flow cytometric analysis indicates that the 2G9 and M5/114.15.2 (Cat. No. 556999, for purified mAb) monoclonal antibodies have comparable reactivity on cells from mice with I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek alloantigens.
Host Species: Rat
Clone: 2G9
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: BALB /c mouse epidermal Langerhans cells
Flow Cytometry
Specifications
| I-A/I-E | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| BALB /c mouse epidermal Langerhans cells | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 2G9 | |
| FITC | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.