Learn More
uvex 9183 i-5 Safety Spectacles
Product Code. 17797679 Shop All Uvex Products
Change view
Click to view available options
Clear; Guard version
Product Code.
17797679 Clear; Guard version
1 options
Product Code. 17797679 Supplier uvex Supplier No. 9183180
  • ONLINE EXCLUSIVE PRICE
    £22.55
    £21.82 / Pack of 1
    Save £0.73
    3% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Reliable protection thanks to an innovative, modern design: with x-tended eyeshield and sideshield, the Uvex i-5 can be relied on to protect the eyes in many situations.
Sophisticated features ensure superior wearer comfort even during long periods of use.
General features
  • Safety spectacles with soft, flexible nose piece
  • Incl. additional frame for even greater wearer comfort and a secure fit
  • The uvex supravision excellence lens coating provides permanent anti-fog protection on the inside and is chemical- and scratch-resistant on the outside
  • Metal-free
  • Frame color: anthracite, blue
  • PC lens: clear
Protective features
  • Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
  • Marking: W 166 FT CE – 2C-1,2 W 1 FTKN CE
  • Reliable UV400 protection
Comfort features
  • Soft, flexible nose pad for maximum wearer comfort and a non-slip fit
  • X-tended eyeshield provides protection against dust, dirt and flying particles
  • X-tended sideshield provides further protection without restricting peripheral vision
  • 3-position inclination allows wearers to achieve a comfortable fit
  • Extendable x-twist side arm technology offers flexible, convenient adjustment to any head shape
  • Soft ear-pieces for a pressure-free, non-slip fit
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Lens Coating Anti-fog, Scratch-resistant
Color Anthracite, Blue
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard