Learn More
uvex i-3 Guard Narrow Safety Glasses
Product Code. 17737689 Shop All Uvex Products
Change view
Click to view available options
Lens Tint: Clear
Product Code.
17737689 Lens Tint: Clear
1 options
Product Code. 17737689 Supplier uvex Supplier No. 9190680
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Uvex i-3 safety glasses can be adapted to any face shape for extreme comfort and a secure fit without compromise.
General features
  • Adjustable, multi-feature premium quality safety spectacle
  • Robust polycarbonate lens with uvex’s proven supravision coating technology – permanent anti-fog and scratch-resistant lens
  • Clip-in frame seal provides extra protection against particles, dirt and dust
  • Narrow version, also available in standard size
  • Metal-free
Safety features
  • Certified in line with EN 166 (personal protective eyewear) and EN 170 (UV protective filter)
  • Marking: W 166 FT CE – 2C-1,2 W1 FTKN CE
  • UV400 – 100% protection against dangerous UVA and UVB radiation up to 400 nm
Comfort features
  • Combination of soft padding around the brow with ergonomically shaped side arms and soft-grip temple ends provide a comfortable, secure fit without pressure
  • Side arms with multi-incline points allow for a high level of individualization to ensure a good fit for all facial shapes and head widths
  • Soft, adjustable nose pad allows spectacle to be adapted to the wearer to deliver a comfortable, non-slip and pressure-free fit
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Chemical-resistant
Lens Coating Anti-fog, Scratch-resistant
Color Anthracite, Blue
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard