Learn More
Hypericin, 98%, Thermo Scientific™
Hypericin, CAS # 548-04-9, is an anthraquinone derivative that shows antidepressant, antiviral, antineoplastic, and immunostimulant activities.
£372.99 - £1046.01
Chemical Identifiers
| CAS | 548-04-9 |
|---|---|
| Molecular Formula | C30H16O8 |
| Molecular Weight (g/mol) | 504.45 |
| MDL Number | MFCD00016683 |
| InChI Key | BTXNYTINYBABQR-UHFFFAOYSA-N |
| Synonym | hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn |
| PubChem CID | 5281051 |
| ChEBI | CHEBI:5835 |
| SMILES | CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11456440
|
Thermo Scientific Alfa Aesar
H26425.MB |
25mg |
£381.00 £372.99 / 25mg Save £8.01 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11466440
|
Thermo Scientific Alfa Aesar
H26425.MC |
100mg |
£1069.00 £1046.01 / 100mg Save £22.99 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
- Hypericin is an anthraquinone derivative found in the yellow flower of Hypericum perforatum (also known as St. John's wort)
- This compound has the ability to inhibit protein kinase C, inducing photosynthesized apoptosis
Application
- It shows antibiotic and antiviral activities
- The antidepressant activity of this compound could be related to its ability to inhibit the neuronal uptake of serotonin, norepinephrine, dopamine, gamma-aminobutyric acid (GABA), and L-glutamate
- Since this compound is able to inhibit the assembly and shedding of virus particles in infected cells, hypericin may also prevent the replication of encapsulated viruses
- It has been described that this compound can also exhibit anti-HIV and anticancer activities
- Hypericin also displays an effective phototoxic activity and is able to trigger apoptotic pathways, resulting in the formation of reactive oxygen species
Chemical Identifiers
| 548-04-9 | |
| 504.45 | |
| BTXNYTINYBABQR-UHFFFAOYSA-N | |
| 5281051 | |
| CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O |
| C30H16O8 | |
| MFCD00016683 | |
| hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn | |
| CHEBI:5835 |
Specifications
| 548-04-9 | |
| MFCD00016683 | |
| Light sensitive | |
| hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn | |
| BTXNYTINYBABQR-UHFFFAOYSA-N | |
| 504.45 | |
| CHEBI:5835 | |
| 98% |
| C30H16O8 | |
| 1917913 | |
| 14,4863 | |
| Soluble in ethanol,dimethyl sulfoxide,methanol,acetone,ethylmethylketone,pyridine and sodium hydroxide. Insoluble in water. | |
| CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O | |
| 5281051 | |
| 504.44 | |
| Hypericin |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
missing translation for 'einecsNumber' : 208-941-0
missing translation for 'rtecsNumber' : SF8472000
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.