missing translation for 'onlineSavingsMsg'
Learn More

Hypericin, 98%

Product Code. 11466440
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466440 100 mg 100mg
11456440 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466440 Supplier Thermo Scientific Alfa Aesar Supplier No. H26425.MC
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hypericin, CAS # 548-04-9, is an anthraquinone derivative that shows antidepressant, antiviral, antineoplastic, and immunostimulant activities. | CAS: 548-04-9 | C30H16O8 | 504.45 g/mol

Inhibitor of protein kinase C; induces photosynthesized apoptosisHypercin acts as an antibiotic and antiviral. It is involved in the inhibition of enzyme dopamine beta-hydroxylase action. It also exhibits an anti-HIV and anticancer drug. Further, it is useful in photodynamic therapy. In addition to this, it serves as an inhibitor of protein kinase C which induces photosynthesized apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hypericin is an anthraquinone derivative found in the yellow flower of Hypericum perforatum (also known as St. John's wort)
  • This compound has the ability to inhibit protein kinase C, inducing photosynthesized apoptosis

Application

  • It shows antibiotic and antiviral activities
  • The antidepressant activity of this compound could be related to its ability to inhibit the neuronal uptake of serotonin, norepinephrine, dopamine, gamma-aminobutyric acid (GABA), and L-glutamate
  • Since this compound is able to inhibit the assembly and shedding of virus particles in infected cells, hypericin may also prevent the replication of encapsulated viruses
  • It has been described that this compound can also exhibit anti-HIV and anticancer activities
  • Hypericin also displays an effective phototoxic activity and is able to trigger apoptotic pathways, resulting in the formation of reactive oxygen species
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-04-9
Molecular Formula C30H16O8
Molecular Weight (g/mol) 504.45
MDL Number MFCD00016683
InChI Key BTXNYTINYBABQR-UHFFFAOYSA-N
Synonym hypericin, hypericum red, hipericina, hypericine, hyperizin, cyclosan, 1,3,4,6,8,13-hexahydroxy-10,11-dimethylphenanthro 1,10,9,8-opqra perylene-7,14-dione, cyclo-werol, hypericum extract, vimrxyn
PubChem CID 5281051
ChEBI CHEBI:5835
SMILES CC1=CC(=C2C3=C1C4=C5C(=C(C=C4C)O)C(=O)C6=C(C=C(C7=C6C5=C3C8=C7C(=CC(=C8C2=O)O)O)O)O)O

Specifications

Melting Point ∼320°C (decomposition)
Quantity 100 mg
Beilstein 1917913
Sensitivity Light sensitive
Merck Index 14,4863
Solubility Information Soluble in ethanol,dimethyl sulfoxide,methanol,acetone,ethylmethylketone,pyridine and sodium hydroxide. Insoluble in water.
Formula Weight 504.44
Percent Purity 98%
Chemical Name or Material Hypericin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.