missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Diethanol, TRC

Product Code. 30401520
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401520 5 mg
30399172 50 mg
30398305 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401520 Supplier Toronto Research Chemicals Supplier No. TRCH9169205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103396-22-1
Molecular Formula C19H28ClN3O2
Molecular Weight (g/mol) 365.9
Synonym 2,2'-((4-((7-Chloro-3-methylquinolin-4-yl)amino)pentyl)azanediyl)diethanol
SMILES CC(NC1=C2C=CC(Cl)=CC2=NC=C1C)CCCN(CCO)CCO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H28ClN3O2/c1-14-13-21-18-12-16(20)5-6-17(18)19(14)22-15(2)4-3-7-23(8-10-24)9-11-25/h5-6,12-13,15,24-25H,3-4,7-11H2,1-2H3,(H,21,22)
Formula Weight 365.187
Chemical Name or Material 3'-Methyl Hydroxychloroquine Diethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.