CAS RN 103396-22-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxychloroquine Diethanol, TRC
CAS: 103396-22-1 Molecular Formula: C19H28ClN3O2 Molecular Weight (g/mol): 365.9 Synonym: 2,2'-((4-((7-Chloro-3-methylquinolin-4-yl)amino)pentyl)azanediyl)diethanol SMILES: CC(NC1=C2C=CC(Cl)=CC2=NC=C1C)CCCN(CCO)CCO