missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Ebastine, TRC

Product Code. 30406060
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406060 5 mg 5mg
30392873 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406060 Supplier Toronto Research Chemicals Supplier No. TRCH9187005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210686-41-2
Molecular Formula C32 H39 N O3
Molecular Weight (g/mol) 485.66
IUPAC Name 4-(4-benzhydryloxypiperidin-1-yl)-1-[4-(1-hydroxy-2-methylpropan-2-yl)phenyl]butan-1-one
SMILES CC(C)(CO)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C32H39NO3/c1-32(2,24-34)28-17-15-25(16-18-28)30(35)14-9-21-33-22-19-29(20-23-33)36-31(26-10-5-3-6-11-26)27-12-7-4-8-13-27/h3-8,10-13,15-18,29,31,34H,9,14,19-24H2,1-2H3
Formula Weight 485.293
Percent Purity >95%
Chemical Name or Material Hydroxy Ebastine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.