CAS RN 210686-41-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Ebastine, TRC
CAS: 210686-41-2 Molecular Formula: C32 H39 N O3 Molecular Weight (g/mol): 485.66 IUPAC Name: 4-(4-benzhydryloxypiperidin-1-yl)-1-[4-(1-hydroxy-2-methylpropan-2-yl)phenyl]butan-1-one SMILES: CC(C)(CO)c1ccc(cc1)C(=O)CCCN2CCC(CC2)OC(c3ccccc3)c4ccccc4