missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone 21-Carboxylic Acid, TRC

Product Code. 30402508
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402508 10 mg 10mg
30403868 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402508 Supplier Toronto Research Chemicals Supplier No. TRCH71470010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57172-48-2
Molecular Formula C21H28O6
Molecular Weight (g/mol) 376.44
Synonym (11beta)-11,17-Dihydroxy-3,20-dioxo-pregn-4-en-21-oic Acid
IUPAC Name 2-[(8S,9S,10R,11R,13S,14R,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoacetic acid
SMILES C[C@]12CCC(=O)C=C1CC[C@H]3[C@H]4CC[C@](O)(C(=O)C(=O)O)[C@@]4(C)C[C@@H](O)[C@H]23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H28O6/c1-19-7-5-12(22)9-11(19)3-4-13-14-6-8-21(27,17(24)18(25)26)20(14,2)10-15(23)16(13)19/h9,13-16,23,27H,3-8,10H2,1-2H3,(H,25,26)/t13-,14+,15+,16+,19-,20-,21-/m0/s1
Formula Weight 376.1886
Chemical Name or Material Hydrocortisone 21-Carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.