CAS RN 57172-48-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydrocortisone 21-Carboxylic Acid, TRC
CAS: 57172-48-2 Molecular Formula: C21H28O6 Molecular Weight (g/mol): 376.44 Synonym: (11beta)-11,17-Dihydroxy-3,20-dioxo-pregn-4-en-21-oic Acid IUPAC Name: 2-[(8S,9S,10R,11R,13S,14R,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoacetic acid SMILES: C[C@]12CCC(=O)C=C1CC[C@H]3[C@H]4CC[C@](O)(C(=O)C(=O)O)[C@@]4(C)C[C@@H](O)[C@H]23