missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™
macron
£38.84 - £110.00
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15538034
|
Avantor Macron
2612-14 |
500 mL | Glass Bottle |
£110.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15587944
|
Avantor Macron
2062-04 |
500 mL | SAFEMOR | ||||||
15558034
|
Avantor Macron
2612-46 |
2.5 L | Glass Bottle |
£88.05
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15548034
|
Avantor Macron
2612-44 |
4 x 2.5 L | Glass | ||||||
Chemische Identifikatoren
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Spezifikation
7647-01-0 | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
Hydrochloric Acid, (HCl 36.5%-38.0%) |
ClH | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
ACS/FCC/NF |
Sicherheit und Handhabung
Signalwort
Gefahrenkategorie
Gefahrenhinweis
Sicherheitshinweis
- Danger
Gefahrenkategorie
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Sicherheitshinweis
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.