missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™

Catalog Number 15538034
104.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 mL
2.5 L
4 x 2.5 L
Packaging:
Glass
Glass Bottle
SAFEMOR
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
Packaging Glass Bottle
Quantity 500 mL
IUPAC Name hydrogen chloride
Formula Weight 36.46
Grade ACS/FCC/NF
Chemical Name or Material Hydrochloric Acid, (HCl 36.5%-38.0%)
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'