missing translation for 'onlineSavingsMsg'
Learn More

Hydrazinecarbothioic Acid O-methyl Ester, TRC

Product Code. 30392055
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392055 25 g
30405524 5 g
30393405 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392055 Supplier Toronto Research Chemicals Supplier No. TRCH69756525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19692-07-0
Molecular Formula C2H6N2OS
Molecular Weight (g/mol) 106.15
Synonym Carbazic Acid Thio-O-methyl Ester, Methoxythiocarbonylhydrazine, Methyl Thiocarbazate, O-Methyl Thiocarbazate, Thiocarbazic Acid O-methyl Ester
IUPAC Name O-methyl N-aminocarbamothioate
SMILES COC(=S)NN

Specifications

Quantity 25 g
InChI Formula InChI=1S/C2H6N2OS/c1-5-2(6)4-3/h3H2,1H3,(H,4,6)
Formula Weight 106.0201
Chemical Name or Material Hydrazinecarbothioic Acid O-methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.