CAS RN 19692-07-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydrazinecarbothioic Acid O-methyl Ester, TRC
CAS: 19692-07-0 Molecular Formula: C2H6N2OS Molecular Weight (g/mol): 106.15 Synonym: Carbazic Acid Thio-O-methyl Ester,Methoxythiocarbonylhydrazine,Methyl Thiocarbazate,O-Methyl Thiocarbazate,Thiocarbazic Acid O-methyl Ester IUPAC Name: O-methyl N-aminocarbamothioate SMILES: COC(=S)NN