Learn More
Invitrogen™ HYD Recombinant Rabbit Monoclonal Antibody (24GB2540)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557827
 View more versions of this product
Product Code. 30276311
  • £172.00 / 20µL
Description
HYD Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
EDD (for E3 identified by Differential Display) is a progestin-regulated gene that was isolated from T-47D human breast cancer cells. Based on sequence homology, EDD appears to be a human homolog of the Drosophila hyperplastic discs (hyd) gene, a tumor suppressor gene that is required for control of imaginal disc growth. EDD contains a HECT domain in the carboxy terminus. HECT domain-containing proteins function as ubiquitin-protein ligases, or E3 enzymes. EDD has been shown to bind to ubiquitin, and like other HECT family proteins, may function as an E3 ubiquitin-protein ligase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYD
Recombinant Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95071, Q62671, Q80TP3
UBR5
A synthesized peptide derived from human EDD (aa 1-54).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2540
Unconjugated
UBR5
CH D1S3362; DD5; E3 identified by differential display; E3 ubiquitin-protein ligase UBR5; E3 ubiquitin-protein ligase, HECT domain-containing 1; ECGF1; Edd; Edd1; EDG1; EDG-1; extraembryonic development; HECT-type E3 ubiquitin transferase UBR5; hHYD; HYD; Hyperplastic discs protein homolog; KIAA0896; Progestin-induced protein; Rat100; S1P receptor 1; S1P receptor Edg-1; S1P1; S1PR1; ubiquitin protein ligase E3 component n-recognin 5; Ubr5
Rabbit
Affinity chromatography
RUO
117060, 51366, 70790
-20°C