Learn More
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels
Brand: Cytiva SH30898.03HI
647.00 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Code : 85
Additional Details : Weight : 0.01000kg
Description
- Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
- Designed for applications where extremely low levels of inherent bovine IgG are required
- Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
- For use in monoclonal antibody production, virus propagation, and immunoassay procedures
Specifications
<-10°C | |
Fetal Bovine Serum | |
Sterile | |
Heat Inactivated | |
Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures | |
≤ 10 mg/dL | |
HyClone™ |
United States | |
500 mL | |
40 nm filtration | |
≤ 10 EU/mL | |
Yes | |
≤ 5 μg/mL |