missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels
£97.50 - £728.00
Specifications
Country of Origin | United States |
---|---|
Product Type | Fetal Bovine Serum |
Sterility | Sterile |
Endotoxin Concentration | ≤ 10 EU/mL |
For Use With (Application) | Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures |
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Processing | Price | Quantity & Availability | ||||
17226916
|
Cytiva
SH30898.01 |
50 mL | None |
£97.50
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
12330143
|
Cytiva
SH30898.02HI |
100 mL | Heat Inactivated |
£180.00
100mL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12350143
|
Cytiva
SH30898.03HI |
500 mL | Heat Inactivated |
£728.00
500mL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
- Designed for applications where extremely low levels of inherent bovine IgG are required
- Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
- For use in monoclonal antibody production, virus propagation, and immunoassay procedures
Specifications
United States | |
Sterile | |
Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures | |
≤ 5 μg/mL |
Fetal Bovine Serum | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
HyClone™ |