missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Enhances cell culture growth, processed via protein G chromatography to substantially reduce immunoglobulin G (IgG) levels
Brand: Cytiva SH30898.02HI
Code : 85
Additional Details : Weight : 0.10000kg
Description
- Processed using protein G chromatography to reduce levels to those required for monoclonal antibody production
- Designed for applications where extremely low levels of inherent bovine IgG are required
- Chromatographically treated to reduce IgG levels to less than 5 μg/mL, while still retaining excellent cell growth properties
- For use in monoclonal antibody production, virus propagation, and immunoassay procedures
Specifications
<-10°C | |
Fetal Bovine Serum | |
Sterile | |
Heat Inactivated | |
Monoclonal Antibody Production, Virus Propagation, and Immunoassay Procedures | |
≤ 10 mg/dL | |
HyClone™ |
United States | |
100 mL | |
40 nm filtration | |
≤ 10 EU/mL | |
Yes | |
≤ 5 μg/mL |