missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronan Synthase 3/HAS3 Antibody (3C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£184.00 - £450.00
Specifications
| Antigen | Hyaluronan Synthase 3/HAS3 |
|---|---|
| Clone | 3C9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161389
|
Novus Biologicals
NBP2-37494 |
0.1 mL |
£450.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609535
|
Novus Biologicals
NBP2-37494-0.025ml |
0.025 mL |
£184.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hyaluronan Synthase 3/HAS3 Monoclonal specifically detects Hyaluronan Synthase 3/HAS3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Hyaluronan Synthase 3/HAS3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 3038 | |
| Purified recombinant fragment of human HAS3 expressed in E. Coli. | |
| Primary | |
| 63 kDa |
| 3C9 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| EC 2.4.1.212, HA synthase 3, hyaluronan synthase 3, Hyaluronate synthase 3, Hyaluronic acid synthase 3 | |
| HAS3 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title