missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hyaluronan Synthase 3/HAS3 Antibody (3C9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37494-0.025ml
This item is not returnable.
View return policy
Description
Hyaluronan Synthase 3/HAS3 Monoclonal specifically detects Hyaluronan Synthase 3/HAS3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Hyaluronan Synthase 3/HAS3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| EC 2.4.1.212, HA synthase 3, hyaluronan synthase 3, Hyaluronate synthase 3, Hyaluronic acid synthase 3 | |
| Mouse | |
| 63 kDa | |
| 0.025 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 3C9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HAS3 | |
| Purified recombinant fragment of human HAS3 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 3038 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction