missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Hyaluronan synthase 1 Antibody (2G5H6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27158-0.1ml
This item is not returnable.
View return policy
Description
Hyaluronan synthase 1 Monoclonal antibody specifically detects Hyaluronan synthase 1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Hyaluronan synthase 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1 | |
| Purified recombinant fragment of human Hyaluronan synthase 1 (AA: 74-399) expressed in E. Coli. | |
| 0.1 mL | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells | |
| 3036 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| ELISA, Western Blot | |
| 2G5H6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu