missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Hyaluronan synthase 1 Antibody (2G5H6), Novus Biologicals™

Mouse Monoclonal Antibody

£217.00 - £389.00

Specifications

Antigen Hyaluronan synthase 1
Clone 2G5H6
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30028894
View Documents
Novus Biologicals™
NBP3-27158-0.025ml
0.025 mL
£217.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30037474
View Documents
Novus Biologicals™
NBP3-27158-0.1ml
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hyaluronan synthase 1 Monoclonal antibody specifically detects Hyaluronan synthase 1 in Human samples. It is validated for ELISA,Western Blot
Specifications

Specifications

Hyaluronan synthase 1
1 mg/mL
ELISA, Western Blot
Unconjugated
Mouse
Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells
PBS
3036
IgG1
Protein G purified
2G5H6
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Purified
RUO
Human
HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1
Purified recombinant fragment of human Hyaluronan synthase 1 (AA: 74-399) expressed in E. Coli.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.