missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Hyaluronan synthase 1 Antibody (2G5H6), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27158-0.025ml

 View more versions of this product

Product Code. 30028894

  • £217.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Hyaluronan synthase 1 Monoclonal antibody specifically detects Hyaluronan synthase 1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hyaluronan synthase 1
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1
Purified recombinant fragment of human Hyaluronan synthase 1 (AA: 74-399) expressed in E. Coli.
0.025 mL
Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells
3036
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
ELISA, Western Blot
2G5H6
Unconjugated
PBS
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.