missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYAL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79911
This item is not returnable.
View return policy
Description
HYAL3 Polyclonal specifically detects HYAL3 in Human samples. It is validated for Western Blot.
Specifications
| HYAL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.2.1.35, Hyal-3, hyaluronidase-3, hyaluronoglucosaminidase 3, Hyaluronoglucosaminidase-3, LUCA14, LuCa-3, LUCA3LUCA-3, Lung carcinoma protein 3, Minna14 | |
| Rabbit | |
| 46 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 86%; Guinea pig: 86%; Rabbit: 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_003540 | |
| HYAL3 | |
| The immunogen for this antibody is HYAL3. Peptide sequence RAAMACSHQRCHGHGRCARRDPGQMEAFLHLWPDGSLGDWKSFSCHCYWG. | |
| Affinity purified | |
| RUO | |
| 8372 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction