Learn More
Hamilton™ HVCX Large Body Ceramic Valves
Product Code. p-7138546
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
4-5 Model
8-5 Model
For Use With:
PSD/2, 3 Syringe Drive Module
PSD/4, 8 Syringe Drive Module
Unit Size:
Each
Product Code. 11573494
Brand: Hamilton™ 650002
Designed to provide superior performance with salt solutions.
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
Specifications
Description 4-5 Model, 4 Ports, Aluminum Housing
For Use With PSD/2, 3 Syringe Drive Module
Material Aluminum
Model 4-5 Model
Type Ceramic Large Body Valve