Learn More
Hamilton™ HVC Standard Ceramic Valves
Product Code. 11593394
Change view
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
3-2 Model
3-5 Model
T 3-2 Model
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Model Unit Size
11593394 Stainless Steel 3-2 Model Each
11543314 Aluminum 3-2 Model Each
11583404 Stainless Steel 3-5 Model Each
11573344 Aluminum 3-5 Model Each
11513484 Aluminum T 3-2 Model Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11593394 Supplier Hamilton™ Supplier No. 676301
  • ONLINE EXCLUSIVE PRICE
    £503.00
    £469.06 / Pack of 1
    Save £33.94
    6% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to provide superior performance with salt solutions.
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
Specifications
Description 3-2 Model, 3 Ports, Stainless Steel Housing
For Use With PSD/2 and 3 Syringe Drive Modules
Material Stainless Steel
Model 3-2 Model
Type Ceramic HV Valve