missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huntingtin Interacting Protein K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £581.00
Specifications
| Antigen | Huntingtin Interacting Protein K |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18128453
|
Novus Biologicals
NBP2-29876-0.025ML |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758253
|
Novus Biologicals
NBP2-29876 |
£581.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Huntingtin Interacting Protein K Polyclonal specifically detects Huntingtin Interacting Protein K in Human, Rat samples. It is validated for Western Blot.Specifications
| Huntingtin Interacting Protein K | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| No buffer with No Preservative | |
| C15orf63, chromosome 15 open reading frame 63, EC 2.1.1.43, EC 2.7.7.6, FLJ20431, HSPC136, Huntingtin yeast partner K, huntingtin-interacting protein K, HYPKHuntingtin interacting protein K | |
| HYPK | |
| IgG | |
| Affinity Purified | |
| Huntingtin-interacting protein 1 (HIP1). By Western Blot the antibody recognizes a band at ∼116 kDa on P2 microsomal fraction from embryonic rat brain. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Rat, Human | |
| O00291 | |
| 25764 | |
| Synthetic peptide. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 116 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title