missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huntingtin Interacting Protein K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29876-0.025ML
This item is not returnable.
View return policy
Description
Huntingtin Interacting Protein K Polyclonal specifically detects Huntingtin Interacting Protein K in Human, Rat samples. It is validated for Western Blot.
Specifications
| Huntingtin Interacting Protein K | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| O00291 | |
| HYPK | |
| Synthetic peptide. | |
| Affinity Purified | |
| RUO | |
| 25764 | |
| Rat, Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| No buffer with No Preservative | |
| C15orf63, chromosome 15 open reading frame 63, EC 2.1.1.43, EC 2.7.7.6, FLJ20431, HSPC136, Huntingtin yeast partner K, huntingtin-interacting protein K, HYPKHuntingtin interacting protein K | |
| Rabbit | |
| 116 kDa | |
| 0.025 mL | |
| Primary | |
| Huntingtin-interacting protein 1 (HIP1). By Western Blot the antibody recognizes a band at ∼116 kDa on P2 microsomal fraction from embryonic rat brain. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction