missing translation for 'onlineSavingsMsg'
Learn More

Huntingtin Antibody (6F2), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen Huntingtin
Clone 6F2
Dilution ELISA, Immunohistochemistry 1:50-1:200
Applications ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30032235
View Documents
Novus Biologicals
NBP3-26212-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30032930
View Documents
Novus Biologicals
NBP3-26212-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Huntingtin Monoclonal antibody specifically detects Huntingtin in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications

Specifications

Huntingtin
ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein
A synthesized peptide derived from Human Huntingtin [UniProt P42858]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
6F2
ELISA, Immunohistochemistry
Unconjugated
Rabbit
Apoptosis, Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
PBS, pH 7.4, 150mM NaCl and 50% glycerol
3064
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.