missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huntingtin Antibody (6F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26212-50ul
This item is not returnable.
View return policy
Description
Huntingtin Monoclonal antibody specifically detects Huntingtin in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Huntingtin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 6F2 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein | |
| A synthesized peptide derived from Human Huntingtin [UniProt P42858] | |
| 50 μL | |
| Apoptosis, Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 3064 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction