missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XCL1/Lymphotactin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£170.00 - £510.00
Specifications
| Antigen | XCL1/Lymphotactin |
|---|---|
| Clone | 109001 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718868
![]() |
R&D Systems
MAB6951-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18143421
|
R&D Systems
MAB6951 |
500 μg |
£510.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| XCL1/Lymphotactin | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| XCL1 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 109001 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P47992 | |
| 6375 | |
| E. coli-derived recombinant human XCL1/Lymphotactin Val22-Gly114 Accession # P47992 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human XCL1/Lymphotactin in ELISAs and Western blots. In sandwich ELISAs, less than 0.15% cross-reactivity was observed with recombinant mouse MCP-5, recombinant human (rh) MIP-3 beta , rhGRO alpha , rhTGF-alpha , rhGRO gamma , rhActivin RII Fc Chimera, rhIGF-II, or rhIL-15. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title