missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XCL1/Lymphotactin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB6951-SP
This item is not returnable.
View return policy
Description
XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| XCL1/Lymphotactin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6375 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 109001 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P47992 | |
| XCL1 | |
| E. coli-derived recombinant human XCL1/Lymphotactin Val22-Gly114 Accession # P47992 | |
| 25 μg | |
| Primary | |
| Detects human XCL1/Lymphotactin in ELISAs and Western blots. In sandwich ELISAs, less than 0.15% cross-reactivity was observed with recombinant mouse MCP-5, recombinant human (rh) MIP-3 beta , rhGRO alpha , rhTGF-alpha , rhGRO gamma , rhActivin RII Fc Chimera, rhIGF-II, or rhIL-15. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction