missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human vWF-A2 (ADAMTS13-cleaved) Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £413.00
Specifications
| Antigen | vWF-A2 |
|---|---|
| Clone | 490628 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797579
![]() |
R&D Systems
MAB27642-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18191801
|
R&D Systems
MAB27642 |
100 μg |
£413.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
vWF-A2 Monoclonal specifically detects vWF-A2 in Human samples. It is validated for Western Blot.Specifications
| vWF-A2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000543 | |
| 7450 | |
| KLH-conjugated human vWF synthetic peptide Asp1596-Tyr1605 Accession # NP_000543 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human vWF-A2 (ADAMTS13-cleaved) in Western blots. This antibody detects a nascent epitope that is generated upon cleavage of recombinant human (rh) vWF-A2 domain with ADAMTS13. It does not detect intact rhvWF-A2 domain. |
| 490628 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| F8VWF, VWD, VWF von Willebrand factor, vWFA2 | |
| VWF | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title