missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human vWF-A2 (ADAMTS13-cleaved) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB27642
This item is not returnable.
View return policy
Description
vWF-A2 Monoclonal specifically detects vWF-A2 in Human samples. It is validated for Western Blot.
Specifications
| vWF-A2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_000543 | |
| VWF | |
| KLH-conjugated human vWF synthetic peptide Asp1596-Tyr1605 Accession # NP_000543 | |
| 100 μg | |
| Primary | |
| Detects human vWF-A2 (ADAMTS13-cleaved) in Western blots. This antibody detects a nascent epitope that is generated upon cleavage of recombinant human (rh) vWF-A2 domain with ADAMTS13. It does not detect intact rhvWF-A2 domain. | |
| Human | |
| Purified |
| Western Blot | |
| 490628 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| F8VWF, VWD, VWF von Willebrand factor, vWFA2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7450 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction