missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF-D Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£157.00 - £413.00
Specifications
| Antigen | VEGF-D |
|---|---|
| Clone | 78902 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711127
![]() |
R&D Systems
MAB2861-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753288
![]() |
R&D Systems
MAB2861-100 |
100 μg |
£413.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789577
![]() |
R&D Systems
MAB2861-500 |
500 μg |
N/A
|
N/A | |||||
Description
VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| VEGF-D | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| O43915 | |
| 2277 | |
| Mouse myeloma cell line NS0-derived recombinant human VEGF-D Phe93-Ser201 Accession # O43915 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human VEGF-D in ELISAs and Western blots. In ELISAs, this antibody shows less than 1% cross-reactivity with recombinant human (rh) VEGF 121 rhVEGF 165 rhVEGF-B 167 rhVEGF-B 186 and rhVEGF-C. In Western blots, this antibody shows 100% cross-reactivity with rmVEGF-D, less than 5% cross-reactivity with rhVEGF 121 rhVEGF 165 rmVEGF 120 and no cross-reactivity with rhVEGF-B 165 rmVEGF-B 186 rhVEGF-B 186 rmVEGF 115 rrVEGF 164 or rhVEGF 206 . |
| 78902 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor | |
| FIGF | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title