missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human VEGF-D Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2861-100
This item is not returnable.
View return policy
Description
VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| VEGF-D | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-fos induced growth factor (vascular endothelial growth factor D), FIGF, vascular endothelial growth factor D, VEGFD, VEGF-DVEGFDc-Fos-induced growth factor | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 2277 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, ELISA | |
| 78902 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O43915 | |
| FIGF | |
| Mouse myeloma cell line NS0-derived recombinant human VEGF-D Phe93-Ser201 Accession # O43915 | |
| 100 μg | |
| Primary | |
| Detects human VEGF-D in ELISAs and Western blots. In ELISAs, this antibody shows less than 1% cross-reactivity with recombinant human (rh) VEGF 121 rhVEGF 165 rhVEGF-B 167 rhVEGF-B 186 and rhVEGF-C. In Western blots, this antibody shows 100% cross-reactivity with rmVEGF-D, less than 5% cross-reactivity with rhVEGF 121 rhVEGF 165 rmVEGF 120 and no cross-reactivity with rhVEGF-B 165 rmVEGF-B 186 rhVEGF-B 186 rmVEGF 115 rrVEGF 164 or rhVEGF 206 . | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction