missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
£168.00 - £511.00
Specifications
| Antigen | ULBP-3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707367
![]() |
R&D Systems
AF1517-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18001032
![]() |
R&D Systems
AF1517 |
100 μg |
£511.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULBP-3 Polyclonal specifically detects ULBP-3 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
| ULBP-3 | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Human | |
| NKG2D ligand 3, RAET1N, UL16 binding protein 3, ULBP3 | |
| ULBP3 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| NP_078794 | |
| 79465 | |
| Mouse myeloma cell line NS0-derived recombinant human ULBP-3 Gly27-Pro216 Accession # NP_078794 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ULBP-3 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) ULBP-2 is observed and less than 1% cross-reactivity with rhULBP-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title