missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1517
This item is not returnable.
View return policy
Description
ULBP-3 Polyclonal specifically detects ULBP-3 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications
| ULBP-3 | |
| Polyclonal | |
| Unconjugated | |
| NP_078794 | |
| ULBP3 | |
| Mouse myeloma cell line NS0-derived recombinant human ULBP-3 Gly27-Pro216 Accession # NP_078794 | |
| 100 μg | |
| Primary | |
| Detects human ULBP-3 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) ULBP-2 is observed and less than 1% cross-reactivity with rhULBP-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| NKG2D ligand 3, RAET1N, UL16 binding protein 3, ULBP3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79465 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction