missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TSP50 Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £353.00
Specifications
| Antigen | TSP50 |
|---|---|
| Clone | 260606 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744249
![]() |
R&D Systems
MAB2430-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18149500
|
R&D Systems
MAB2430 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSP50 Monoclonal specifically detects TSP50 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| TSP50 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Human | |
| Cancer/testis antigen 20, CT20probable threonine protease PRSS50, EC 3.4.25.-, protease, serine, 50, Serine protease 50, testes-specific protease 50, Testis-specific protease-like protein 50, TSP50testes specific protease 50 | |
| PRSS50 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 260606 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 29122 | |
| Mouse myeloma cell line NS0-derived recombinant human TSP50 Ala40-Ala363 (predicted) | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TSP50 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactvity with recombinant mouse TSP50 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title